Thermo Scientific Chemicals

3-Amino-2-chloropyridine, 96%, Thermo Scientific Chemicals

Catalog number: 103420500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Amino-2-chloropyridine, 96%, Thermo Scientific Chemicals

Catalog number: 103420500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 103420500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6298-19-7
IUPAC Name
2-chloropyridin-3-amine
Molecular Formula
C5H5ClN2
InChI Key
MEQBJJUWDCYIAB-UHFFFAOYSA-N
SMILES
NC1=CC=CN=C1Cl
Appearance (Form)
Crystalline powder or crystals and/or chunks
Melting point
76°C to 81°C
Appearance (Color)
Off-white to yellow to pink
Infrared spectrum
Conforms
HPLC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text