Thermo Scientific Chemicals

1-Amino-1-cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 103470050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Amino-1-cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 103470050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 103470050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52-52-8
IUPAC Name
1-aminocyclopentane-1-carboxylic acid
Molecular Formula
C6H11NO2
InChI Key
NILQLFBWTXNUOE-UHFFFAOYSA-N
SMILES
NC1(CCCC1)C(O)=O
Appearance (Color)
White to beige
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or flakes
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text