2-Amino-4,6-dimethylpyridine, 99%, Thermo Scientific Chemicals
2-Amino-4,6-dimethylpyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4,6-dimethylpyridine, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 103580050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5407-87-4
IUPAC Name4,6-dimethylpyridin-2-amine
Molecular FormulaC7H10N2
InChI KeyBRBUBVKGJRPRRD-UHFFFAOYSA-N
SMILESCC1=CC(C)=NC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point64°C to 70°C
Appearance (Form)Crystalline powder or solid
GC>=98.5 %
Appearance (Color)White to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only