2-(2-Aminoethoxy)ethanol, 98%, Thermo Scientific Chemicals
2-(2-Aminoethoxy)ethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(2-Aminoethoxy)ethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 103631000
Price (USD)/ Each
43.65
Online exclusive
51.60 
Save 7.95 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
51.60 
Save 7.95 (15%)
Add to cart
2-(2-Aminoethoxy)ethanol, 98%, Thermo Scientific Chemicals
Catalog number103631000
Price (USD)/ Each
43.65
Online exclusive
51.60 
Save 7.95 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-(2-aminoethoxy)ethan-1-ol
Molecular FormulaC4H11NO2
InChI KeyGIAFURWZWWWBQT-UHFFFAOYSA-N
SMILESNCCOCCO
Molecular Weight (g/mol)105.14
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4570 to 1.4610 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only