N-(2-Aminoethyl)pyrrolidine, 99%, Thermo Scientific Chemicals
N-(2-Aminoethyl)pyrrolidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(2-Aminoethyl)pyrrolidine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
100 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 103700250
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
Add to cart
N-(2-Aminoethyl)pyrrolidine, 99%, Thermo Scientific Chemicals
Catalog number103700250
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7154-73-6
IUPAC Name2-(pyrrolidin-1-yl)ethan-1-amine
Molecular FormulaC6H14N2
InChI KeyWRXNJTBODVGDRY-UHFFFAOYSA-N
SMILESNCCN1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4677 to 1.4697 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only