Thermo Scientific Chemicals

N-(2-Aminoethyl)pyrrolidine, 99%, Thermo Scientific Chemicals

Catalog number: 103701000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

N-(2-Aminoethyl)pyrrolidine, 99%, Thermo Scientific Chemicals

Catalog number: 103701000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 103701000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7154-73-6
IUPAC Name
2-(pyrrolidin-1-yl)ethan-1-amine
Molecular Formula
C6H14N2
InChI Key
WRXNJTBODVGDRY-UHFFFAOYSA-N
SMILES
NCCN1CCCC1
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4677 to 1.4697 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text