Thermo Scientific Chemicals

2-Amino-4-hydroxy-6-methylpyrimidine, 99%, Thermo Scientific Chemicals

Catalog number: 103831000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Amino-4-hydroxy-6-methylpyrimidine, 99%, Thermo Scientific Chemicals

Catalog number: 103831000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 103831000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3977-29-5
IUPAC Name
2-amino-6-methyl-1,4-dihydropyrimidin-4-one
Molecular Formula
C5H7N3O
InChI Key
KWXIPEYKZKIAKR-UHFFFAOYSA-N
SMILES
CC1=CC(=O)N=C(N)N1
Appearance (Color)
White to cream
Infrared spectrum
Conforms
Appearance (Form)
Powder
Titration with HClO4
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text