2-Aminoindan hydrochloride, 98%, Thermo Scientific Chemicals
2-Aminoindan hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminoindan hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 103860010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2338-18-3
IUPAC Name2,3-dihydro-1H-inden-2-amine hydrochloridyl
Molecular FormulaC9H11ClN
InChI KeyDLYRFJDJKGGVKT-UHFFFAOYSA-N
SMILES[Cl].NC1CC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only