2-Aminoindan hydrochloride, 98%, Thermo Scientific Chemicals
2-Aminoindan hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminoindan hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number 103860250
Price (USD)/ Each
395.65
Online exclusive
439.00 
Save 43.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
395.65
Online exclusive
439.00 
Save 43.35 (10%)
Add to cart
2-Aminoindan hydrochloride, 98%, Thermo Scientific Chemicals
Catalog number103860250
Price (USD)/ Each
395.65
Online exclusive
439.00 
Save 43.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,3-dihydro-1H-inden-2-amine hydrochloridyl
Molecular FormulaC9H11ClN
InChI KeyDLYRFJDJKGGVKT-UHFFFAOYSA-N
SMILES[Cl].NC1CC2=CC=CC=C2C1
Molecular Weight (g/mol)168.64
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only