Thermo Scientific Chemicals

2-Amino-2-methyl-1,3-propanediol, 99%, Thermo Scientific Chemicals

Catalog number: 104050025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

2-Amino-2-methyl-1,3-propanediol, 99%, Thermo Scientific Chemicals

Catalog number: 104050025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 104050025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
115-69-5
IUPAC Name
2-amino-2-methylpropane-1,3-diol
Molecular Formula
C4H11NO2
InChI Key
UXFQFBNBSPQBJW-UHFFFAOYSA-N
SMILES
CC(N)(CO)CO
Titration with HClO4
>=98.5 % (on anhydrous substance)
Water
=<0.5 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text