2-Amino-2-methyl-1,3-propanediol, 99%, Thermo Scientific Chemicals
2-Amino-2-methyl-1,3-propanediol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-2-methyl-1,3-propanediol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 104055000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-amino-2-methylpropane-1,3-diol
Molecular FormulaC4H11NO2
InChI KeyUXFQFBNBSPQBJW-UHFFFAOYSA-N
SMILESCC(N)(CO)CO
Molecular Weight (g/mol)105.14
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 % (on anhydrous substance)
Water=<0.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only