2-Amino-4-methylpyrimidine, 97%, Thermo Scientific Chemicals
2-Amino-4-methylpyrimidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4-methylpyrimidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 104090100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS108-52-1
IUPAC Name4-methylpyrimidin-2-amine
Molecular FormulaC5H7N3
InChI KeyGHCFWKFREBNSPC-UHFFFAOYSA-N
SMILESCC1=CC=NC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
Melting point155°C to 160°C
Appearance (Color)Off-white to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only