2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminophenol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 104261000
Price (USD)/ Each
35.65
Online exclusive
39.60 
Save 3.95 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
35.65
Online exclusive
39.60 
Save 3.95 (10%)
Add to cart
2-Aminophenol, 99%, Thermo Scientific Chemicals
Catalog number104261000
Price (USD)/ Each
35.65
Online exclusive
39.60 
Save 3.95 (10%)
-
Add to cart
Chemical Identifiers
CAS95-55-6
IUPAC Name2-aminophenol
Molecular FormulaC6H7NO
InChI KeyCDAWCLOXVUBKRW-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
GC>=98.5 %
Appearance (Color)White to brown
Melting point172°C to 177°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only