Thermo Scientific Chemicals

3-Aminophenylboronic acid hemisulfate, 98%, Thermo Scientific Chemicals

Catalog number: 104300050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Aminophenylboronic acid hemisulfate, 98%, Thermo Scientific Chemicals

Catalog number: 104300050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 104300050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
66472-86-4
IUPAC Name
bis((3-aminophenyl)boronic acid); sulfuric acid
Molecular Formula
C12H18B2N2O8S
InChI Key
UKTAURVTSWDIQR-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.NC1=CC=CC(=C1)B(O)O.NC1=CC=CC(=C1)B(O)O
Appearance (Color)
White to beige to grey to light brown
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
Titration with NaOH
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text