4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminophenyl ether, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 104331000
Price (USD)/ Each
64.40
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
64.40
Add to cart
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
Catalog number104331000
Price (USD)/ Each
64.40
-
Add to cart
Chemical Identifiers
CAS101-80-4
IUPAC Name4-(4-aminophenoxy)aniline
Molecular FormulaC12H12N2O
InChI KeyHLBLWEWZXPIGSM-UHFFFAOYSA-N
SMILESNC1=CC=C(OC2=CC=C(N)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Appearance (Color)Beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point189°C to 193°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only