Thermo Scientific Chemicals

4-Aminophenyl ether, 98%, Thermo Scientific Chemicals

Catalog number: 104335000
500 g, Each
Thermo Scientific Chemicals

4-Aminophenyl ether, 98%, Thermo Scientific Chemicals

Catalog number: 104335000
500 g, Each
Quantity
Catalog number: 104335000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101-80-4
IUPAC Name
4-(4-aminophenoxy)aniline
Molecular Formula
C12H12N2O
InChI Key
HLBLWEWZXPIGSM-UHFFFAOYSA-N
SMILES
NC1=CC=C(OC2=CC=C(N)C=C2)C=C1
Titration with HClO4
>=97.5 %
Appearance (Color)
Beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
189°C to 193°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text