Thermo Scientific Chemicals

1-Aminopiperidine, 97%, Thermo Scientific Chemicals

Catalog number: 104410250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Aminopiperidine, 97%, Thermo Scientific Chemicals

Catalog number: 104410250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 104410250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2213-43-6
IUPAC Name
piperidin-1-amine
Molecular Formula
C5H12N2
InChI Key
LWMPFIOTEAXAGV-UHFFFAOYSA-N
SMILES
NN1CCCCC1
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4735 to 1.4775 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text