Thermo Scientific Chemicals

3-Amino-1,2-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: 104422500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Amino-1,2-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: 104422500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 104422500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
616-30-8
IUPAC Name
3-aminopropane-1,2-diol
Molecular Formula
C3H9NO2
InChI Key
KQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILES
NCC(O)CO
Titration with HClO4
>=97.5 %
Water
=<0.5 % (K.F.)
Refractive index
1.4900 to 1.4940 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Viscous liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text