3-Amino-1-propanesulfonic acid, 95%, Thermo Scientific Chemicals
3-Amino-1-propanesulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-1-propanesulfonic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 104430050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3687-18-1
IUPAC Name3-aminopropane-1-sulfonic acid
Molecular FormulaC3H9NO3S
InChI KeySNKZJIOFVMKAOJ-UHFFFAOYSA-N
SMILESNCCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light blue
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=94.0 %
Melting point>=270°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only