3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-1-propanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 104465000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS156-87-6
IUPAC Name3-aminopropan-1-ol
Molecular FormulaC3H9NO
InChI KeyWUGQZFFCHPXWKQ-UHFFFAOYSA-N
SMILESNCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4595 to 1.4615 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only