Thermo Scientific Chemicals

3-Aminopropionitrile fumarate, 99+%, Thermo Scientific Chemicals

Catalog number: 104470250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

3-Aminopropionitrile fumarate, 99+%, Thermo Scientific Chemicals

Catalog number: 104470250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 104470250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2079-89-2
IUPAC Name
(2E)-but-2-enedioic acid; bis(3-aminopropanenitrile)
Molecular Formula
C10H16N4O4
InChI Key
NYMXYZMHOZAPHQ-SEPHDYHBSA-N
SMILES
NCCC#N.NCCC#N.OC(=O)\C=C\C(O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with HClO4
>=99.0 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text