3-Aminopropionitrile fumarate, 99+%, Thermo Scientific Chemicals
3-Aminopropionitrile fumarate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminopropionitrile fumarate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 104471000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2079-89-2
IUPAC Name(2E)-but-2-enedioic acid; bis(3-aminopropanenitrile)
Molecular FormulaC10H16N4O4
InChI KeyNYMXYZMHOZAPHQ-SEPHDYHBSA-N
SMILESNCCC#N.NCCC#N.OC(=O)\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 %
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only