Thermo Scientific Chemicals

Aminopyrazine, 99+%, Thermo Scientific Chemicals

Catalog number: 104490100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Aminopyrazine, 99+%, Thermo Scientific Chemicals

Catalog number: 104490100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 104490100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5049-61-6
IUPAC Name
pyrazin-2-amine
Molecular Formula
C4H5N3
InChI Key
XFTQRUTUGRCSGO-UHFFFAOYSA-N
SMILES
NC1=CN=CC=N1
Appearance (Color)
Light yellow to beige
Infrared spectrum
Conforms
Titration with HClO4
>=99.0 %
Appearance (Form)
Crystalline powder
Melting point
117°C to 120°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text