3-Aminopyrazine-2-carboxylic acid, 99+%, Thermo Scientific Chemicals
3-Aminopyrazine-2-carboxylic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminopyrazine-2-carboxylic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 104500050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5424-01-1
IUPAC Name3-aminopyrazine-2-carboxylic acid
Molecular FormulaC5H5N3O2
InChI KeyZAGZIOYVEIDDJA-UHFFFAOYSA-N
SMILESNC1=NC=CN=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=99.0 %
Appearance (Color)Yellow-green to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only