Thermo Scientific Chemicals

1-Aminopyrene, 97%, Thermo Scientific Chemicals

Catalog number: 104530050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Aminopyrene, 97%, Thermo Scientific Chemicals

Catalog number: 104530050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 104530050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1606-67-3
IUPAC Name
pyren-1-amine
Molecular Formula
C16H11N
InChI Key
YZVWKHVRBDQPMQ-UHFFFAOYSA-N
SMILES
NC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Appearance (Color)
Yellow to green
Infrared spectrum
Conforms
Titration with HClO4
>=96.0 %
Appearance (Form)
Powder
Melting point
114°C to 118°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text