Thermo Scientific Chemicals

2-Aminopyridine, 99+%, Thermo Scientific Chemicals

Catalog number: 104545000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Aminopyridine, 99+%, Thermo Scientific Chemicals

Catalog number: 104545000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 104545000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
504-29-0
IUPAC Name
pyridin-2-amine
Molecular Formula
C5H6N2
InChI Key
ICSNLGPSRYBMBD-UHFFFAOYSA-N
SMILES
NC1=CC=CC=N1
Melting point
55°C to 60°C
GC
>=99.0 %
Appearance (Color)
White to yellow to light beige
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals or flakes and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text