4-Aminosalicylic acid, 99%, Thermo Scientific Chemicals
4-Aminosalicylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminosalicylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Catalog number 104621000
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
Add to cart
4-Aminosalicylic acid, 99%, Thermo Scientific Chemicals
Catalog number104621000
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
-
Add to cart
Chemical Identifiers
CAS65-49-6
IUPAC Name4-amino-2-hydroxybenzoic acid
Molecular FormulaC7H7NO3
InChI KeyWUBBRNOQWQTFEX-UHFFFAOYSA-N
SMILESNC1=CC=C(C(O)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Titration with NaOH>=98.5 %
Appearance (Color)Beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only