Thermo Scientific Chemicals

4-Aminosalicylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 104625000
500 g, Each
Thermo Scientific Chemicals

4-Aminosalicylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 104625000
500 g, Each
Quantity
Catalog number: 104625000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65-49-6
IUPAC Name
4-amino-2-hydroxybenzoic acid
Molecular Formula
C7H7NO3
InChI Key
WUBBRNOQWQTFEX-UHFFFAOYSA-N
SMILES
NC1=CC=C(C(O)=O)C(O)=C1
Infrared spectrum
Conforms
Appearance (Form)
Powder
Titration with NaOH
>=98.5 %
Appearance (Color)
Beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text