Thermo Scientific Chemicals

11-Aminoundecanoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 104711000
100 g, Each
Thermo Scientific Chemicals

11-Aminoundecanoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 104711000
100 g, Each
Quantity
Catalog number: 104711000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2432-99-7
IUPAC Name
11-aminoundecanoic acid
Molecular Formula
C11H23NO2
InChI Key
GUOSQNAUYHMCRU-UHFFFAOYSA-N
SMILES
NCCCCCCCCCCC(O)=O
Appearance (Color)
White
Melting point
187°C to 190°C
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text