o-Anisidine, 99+%, Thermo Scientific Chemicals
o-Anisidine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Anisidine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
250 mL
Packaging:
Glass bottle
Catalog number 104810010
Price (USD)/ Each
117.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
138.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
117.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
138.00 
Save 20.35 (15%)
Add to cart
o-Anisidine, 99+%, Thermo Scientific Chemicals
Catalog number104810010
Price (USD)/ Each
117.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
138.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyVMPITZXILSNTON-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1N
Molecular Weight (g/mol)123.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.0 %
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
Refractive index1.5730 to 1.5755 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only