m-Anisidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
m-Anisidine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
250 mL
1 L
Catalog number 104821000
Price (USD)/ Each
101.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
101.00
Add to cart
m-Anisidine, 99%, Thermo Scientific Chemicals
Catalog number104821000
Price (USD)/ Each
101.00
-
Add to cart
Chemical Identifiers
CAS536-90-3
IUPAC Name3-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyNCBZRJODKRCREW-UHFFFAOYSA-N
SMILESCOC1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5800 to 1.5820 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only