Thermo Scientific Chemicals

Anisoin, 95%, Thermo Scientific Chemicals

Catalog number: 104841000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Anisoin, 95%, Thermo Scientific Chemicals

Catalog number: 104841000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 104841000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-52-8
IUPAC Name
2-hydroxy-1,2-bis(4-methoxyphenyl)ethan-1-one
Molecular Formula
C16H16O4
InChI Key
LRRQSCPPOIUNGX-UHFFFAOYNA-N
SMILES
COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
HPLC
>=94.0 %
Appearance (Form)
Powder
Melting point
108°C to 115°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text