Anthracene-9-carboxylic acid, 99%, Thermo Scientific Chemicals
Anthracene-9-carboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Anthracene-9-carboxylic acid, 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 104880100
Price (USD)/ Each
90.50
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
90.50
Add to cart
Anthracene-9-carboxylic acid, 99%, Thermo Scientific Chemicals
Catalog number104880100
Price (USD)/ Each
90.50
-
Add to cart
Chemical Identifiers
IUPAC Nameanthracene-9-carboxylic acid
Molecular FormulaC15H10O2
InChI KeyXGWFJBFNAQHLEF-UHFFFAOYSA-N
SMILESOC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
Molecular Weight (g/mol)222.24
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow
Melting point214°C to 218°C
Appearance (Form)Powder
Titration with TBAH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only