TrackIt™ 1 Kb Plus DNA Ladder
This product was upgraded on September 15, 2017. Learn more at thermofisher.com/ladderupgrade17.
TrackIt™ 1 Kb Plus DNA Ladder
Invitrogen™

TrackIt™ 1 Kb Plus DNA Ladder

Green features
Invitrogen TrackIt 1 Kb Plus DNA Ladder is ready to use and designed for sizing and approximate quantification of double-strandedRead more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
Have Questions?
Catalog NumberQuantity
10488085Promo Image50 μg
Catalog number 10488085
Price (USD)
103.65
Online Exclusive
112.00
Save 8.35 (7%)
Each
In stock
Add to cart
Quantity:
50 μg
Request bulk or custom format
Price (USD)
103.65
Online Exclusive
112.00
Save 8.35 (7%)
Each
Add to cart

Invitrogen TrackIt 1 Kb Plus DNA Ladder is ready to use and designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 15,000 bp. 1 Kb Plus DNA Ladder consists of 18 individual chromatography-purified DNA fragments and has a reference band at 1,500 bp for easy orientation.

TrackIt 1 Kb DNA Ladder is ideal for separation on 0.8–1% agarose gels.

Highlights of TrackIt 1 Kb Plus DNA Ladder:

  • Sharp, clear bands—chromatography purified fragments for consistent and reliable results and stable at room temperature up to six months
  • Ready-to-use—premixed with 6X TrackIt Cyan/Orange Loading Buffer for tracking of DNA migration
  • Precise—an exact amount of DNA in each band

Product use

The double-stranded DNA ladder can be visualized on 0.8–1% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.1 μg/μL
Gel CompatibilityAgarose gel
Green FeaturesSustainable packaging
No. of Reactions100 Applications
Product LineTrackIt™
Product TypeDNA Ladder
Quantity50 μg
Ready to LoadYes
Sample Loading Volume1 mL
Shelf Life24 Months when stored at room temperature.
Shipping ConditionApproved for shipment at Room Temperature or on Dry Ice
TechnologyIndividual chromatography-purified DNA fragments
Volume (Metric)500 μL
Gel TypeAgarose
Size Range100 to 15000 bp
Unit SizeEach
Contents & Storage
• 500 μL TrackIt 1 Kb Plus DNA Ladder
• 1 mL 6X TrackIt Cyan/Orange Loading Buffer

Store at room temperature or at 4°C for up to 24 months. For longer periods, store at -20°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection