1x1 image pixel for data collection
E-Gel™ 1 Kb Plus DNA Ladder
This product was upgraded on September 15, 2017. Learn more at thermofisher.com/ladderupgrade17.
E-Gel™ 1 Kb Plus DNA Ladder
Invitrogen™

E-Gel™ 1 Kb Plus DNA Ladder

Green features
Invitrogen E-Gel 1 Kb Plus DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the rangeRead more
Have Questions?
Catalog NumberQuantity
10488090100 applications
Catalog number 10488090
Price (USD)
237.00
Special offer
Online exclusive
Ends: 21-Jul-2025
266.00
Save 29.00 (11%)
Each
Add to cart
Quantity:
100 applications
Price (USD)
237.00
Special offer
Online exclusive
Ends: 21-Jul-2025
266.00
Save 29.00 (11%)
Each
Add to cart
Invitrogen E-Gel 1 Kb Plus DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 15,000 bp. E-Gel 1 Kb Plus DNA Ladder consists of 18 individual chromatography-purified DNA fragments and has a reference band at 1,500 bp for easy orientation.

E-Gel 1 Kb Plus DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.

Highlights of E-Gel 1 Kb Plus DNA Ladder:
Performance—optimal for use on E-Gel agarose gels
Sharp, clear bands—chromatography purified fragments for consistent and reliable results and stable at room temperature up to six months
Ready-to-use—premixed with loading buffer
Convenient—provided with 1X E-Gel Sample Loading Buffer
Precise—an exact amount of DNA in each band

Product use
The double-stranded DNA ladder can be visualized on 0.8–1% E-Gel agarose gels. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.025 μg/μL
Gel CompatibilityE-Gel™ agarose gels
Green FeaturesSustainable packaging
Nucleic Acid StructureLinear DNA
Product TypeDNA Ladder
Quantity100 applications
Ready to LoadYes
Sample Loading Volume1 mL
Shelf Life6 Months
Shipping ConditionApproved for shipment at Room Temperature or on Dry Ice
TechnologyIndividual chromatography-purified DNA fragments
Volume (Metric)2 x 1000 μL
Gel TypeAgarose
Size Range100 to 15000 bp
Unit SizeEach
Contents & Storage
• 2 x 1 mL E-Gel 1 Kb Plus DNA Ladder
• 1 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods, store at -20°C.

Frequently asked questions (FAQs)

Can I know the sequences of Invitrogen DNA ladders?

Sequences of Invitrogen DNA and RNA ladders are proprietary.

Are Invitrogen DNA ladders composed of linear or circular/supercoiled DNA?

Invitrogen DNA ladders contain linear dsDNA fragments.

Are Invitrogen DNA ladders composed of single-stranded or double-stranded DNA fragments?

Invitrogen DNA ladders are composed of double-stranded DNA fragments only.

I'm seeing anomalous migration of my DNA ladder. What happened?

This can happen if the marker was heated. Please ensure that the ladders are not heated before use.

What is the difference between the TrackIt DNA Ladders and other DNA ladders?

TrackIt DNA Ladders contain two tracking dyes in the sample buffers, which serve as visual markers for tracking electrophoresis progress through the gel, and also to indicate when maximum resolution is achieved. The tracking dyes should not obscure your visualization of DNA bands in the ladder, as the dyes run outside the limits of most DNA bands in the ladder.

TrackIt Cyan/Orange Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Orange G. We recommend TrackIt Cyan/Orange Loading Buffer for DNA fragments between 10 bp and 1 kb.

The TrackIt Cyan/Yellow Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Tartrazine. We recommend TrackIt Cyan/Yellow Loading Buffer for DNA fragments between 100 bp and 10 kb. The molecular weights are Xylene Cyanol FF, 638.6; Orange G, 452.4; Tartrazine, 534.4.

Note: The TrackIt DNA Ladders are not recommended for use with polyacrylamide gels and are not designed for quantitation.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection