Thermo Scientific Chemicals

2-Aminobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: 104921000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Aminobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: 104921000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 104921000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1885-29-6
IUPAC Name
2-aminobenzonitrile
Molecular Formula
C7H6N2
InChI Key
HLCPWBZNUKCSBN-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1C#N
Appearance (Color)
Off-white to yellow to beige to brown
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals or flakes and/or chunks
Melting point
46°C to 51°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text