Anthraquinone, 98%, Thermo Scientific Chemicals
Anthraquinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Anthraquinone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 104930010
Price (USD)/ Each
163.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
192.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
163.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
192.00 
Save 28.35 (15%)
Add to cart
Anthraquinone, 98%, Thermo Scientific Chemicals
Catalog number104930010
Price (USD)/ Each
163.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
192.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS84-65-1
IUPAC Name9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O2
InChI KeyRZVHIXYEVGDQDX-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)Yellow-green to tan to khaki
Melting point283°C to 288°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only