Anthraquinone, 98%
Anthraquinone, 98%
Anthraquinone, 98%
Anthraquinone, 98%
Thermo Scientific Chemicals

Anthraquinone, 98%

CAS: 84-65-1 | C14H8O2 | 208.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 104932500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS84-65-1
IUPAC Name9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O2
InChI KeyRZVHIXYEVGDQDX-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)Yellow-green to tan to khaki
Melting point283°C to 288°C
Infrared spectrumConforms
Ask our AI about this Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0461617Certificate of AnalysisJun 11, 2024104930010, 104932500, 104930500
A0355824Certificate of AnalysisAug 25, 2023104930010, 104932500, 104930500
A0452671Certificate of AnalysisMay 05, 2023104930010, 104932500, 104930500
2545074HCertificate of AnalysisMar 07, 2023104930010, 104932500, 104930500
A0414099Certificate of AnalysisDec 13, 2022104930010, 104932500, 104930500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link