Thermo Scientific Chemicals

Anthrone, ACS reagent, Thermo Scientific Chemicals

Catalog number: 104961000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Anthrone, ACS reagent, Thermo Scientific Chemicals

Catalog number: 104961000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 104961000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-44-8
IUPAC Name
9,10-dihydroanthracen-9-one
Molecular Formula
C14H10O
InChI Key
RJGDLRCDCYRQOQ-UHFFFAOYSA-N
SMILES
O=C1C2=CC=CC=C2CC2=CC=CC=C12
Appearance (Color)
Yellow
Melting point
152°C to 157°C
Absorbance
=<0.75 (425 nm, 1 cm cell)
Infrared spectrum
Conforms
Absorbance
=<0.045 (620 nm, 1 cm cell)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text