Thermo Scientific Chemicals

L(+)-Arginine, 98+%, Thermo Scientific Chemicals

Catalog number: 104990100
10 kg, Plastic Drum, Each
Thermo Scientific Chemicals

L(+)-Arginine, 98+%, Thermo Scientific Chemicals

Catalog number: 104990100
10 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 104990100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
74-79-3
IUPAC Name
2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Formula
C6H14N4O2
InChI Key
ODKSFYDXXFIFQN-UHFFFAOYNA-N
SMILES
NC(CCCN=C(N)N)C(O)=O
Appearance (Form)
Crystalline powder or crystals
Loss on drying
=<0.5 % (1 g, 105°C)
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Heavy metals (as Pb)
=<15 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text