Thermo Scientific Chemicals

L(+)-Aspartic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 105045000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

L(+)-Aspartic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 105045000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 105045000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-84-8
IUPAC Name
2-aminobutanedioic acid
Molecular Formula
C4H7NO4
InChI Key
CKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILES
NC(CC(O)=O)C(O)=O
Appearance (Color)
White to almost white
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Titration with HClO4
98.0 to 102.0 % (on dried substance)
Loss on drying
=<0.5 % (105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text