Thermo Scientific Chemicals

7-Azaindole, 98%, Thermo Scientific Chemicals

Catalog number: 105100050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

7-Azaindole, 98%, Thermo Scientific Chemicals

Catalog number: 105100050
5 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 105100050
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
271-63-6
IUPAC Name
1H-pyrrolo[2,3-b]pyridine
Molecular Formula
C7H6N2
InChI Key
MVXVYAKCVDQRLW-UHFFFAOYSA-N
SMILES
N1C=CC2=CC=CN=C12
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
103°C to 107°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Serious Eye Damage/Eye Irritation
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H319 - Causes serious eye irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing