(S)-(-)-2-Azetidinecarboxylic acid, 99+%, Thermo Scientific Chemicals
(S)-(-)-2-Azetidinecarboxylic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(-)-2-Azetidinecarboxylic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 105140050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2133-34-8
IUPAC Name(2S)-azetidine-2-carboxylic acid
Molecular FormulaC4H7NO2
InChI KeyIADUEWIQBXOCDZ-VKHMYHEASA-N
SMILESOC(=O)[C@@H]1CCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
TLC>=99.0 %
Specific optical rotation-123° to -127° (25°C, 589 nm) (c=1, water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only