Azulene, 99%
Azulene, 99%
Azulene, 99%
Thermo Scientific Chemicals

Azulene, 99%

CAS: 275-51-4 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number 105181000
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS275-51-4
IUPAC Nameazulene
Molecular FormulaC10H8
InChI KeyCUFNKYGDVFVPHO-UHFFFAOYSA-N
SMILESC1=CC2=CC=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point99°C to 101°C
GC>=98.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471198Certificate of AnalysisMay 17, 2025105180010, 105181000
A0466501Certificate of AnalysisOct 04, 2024105180010, 105181000
A0462222Certificate of AnalysisMay 07, 2024105180010, 105181000
A0458838Certificate of AnalysisJan 05, 2024105180010, 105181000
A0452122Certificate of AnalysisMay 02, 2023105180010, 105181000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection