Thermo Scientific Chemicals

Benzamidine Hydrochloride Hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 105240250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Benzamidine Hydrochloride Hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 105240250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 105240250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
206752-36-5
IUPAC Name
benzenecarboximidamide hydrochloride
Molecular Formula
C7H9ClN2
InChI Key
LZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILES
Cl.NC(=N)C1=CC=CC=C1
Titration Argentometric
97.5 to 102.5 % (on dry substance)
Infrared spectrum
Conforms
Appearance (Form)
Powder
Water
=<19 % (K.F.)
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text