Thermo Scientific Chemicals

1,2-Benzanthracene, 99%, Thermo Scientific Chemicals

Catalog number: 105252500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Benzanthracene, 99%, Thermo Scientific Chemicals

Catalog number: 105252500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 105252500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56-55-3
IUPAC Name
tetraphene
Molecular Formula
C18H12
InChI Key
DXBHBZVCASKNBY-UHFFFAOYSA-N
SMILES
C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1
Appearance (Color)
Beige to yellow-green
Appearance (Form)
Crystalline powder
Melting point
158°C to 161°C
GC
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text