Thermo Scientific Chemicals

2,3-Benzanthracene, 98%, Thermo Scientific Chemicals

Catalog number: 105265000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

2,3-Benzanthracene, 98%, Thermo Scientific Chemicals

Catalog number: 105265000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 105265000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92-24-0
IUPAC Name
tetracene
Molecular Formula
C18H12
InChI Key
IFLREYGFSNHWGE-UHFFFAOYSA-N
SMILES
C1=CC2=CC3=CC4=CC=CC=C4C=C3C=C2C=C1
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
HPLC
>=97.5 %
Appearance (Color)
Orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text