1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 105315000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebenzene-1,2,4,5-tetracarboxylic acid
Molecular FormulaC10H6O8
InChI KeyCYIDZMCFTVVTJO-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(C(O)=O)=C(C=C1C(O)=O)C(O)=O
Molecular Weight (g/mol)254.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH>=95 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only