Thermo Scientific Chemicals

1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: 105320025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: 105320025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 105320025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
89-32-7
IUPAC Name
5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
Molecular Formula
C10H2O6
InChI Key
ANSXAPJVJOKRDJ-UHFFFAOYSA-N
SMILES
O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O
Infrared spectrum
Conforms
Titration excess NaOH
>=98.5 %
Appearance (Color)
White to light beige
Appearance (Form)
Powder
Melting point
283°C to 287°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text