1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 105325000
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
Add to cart
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
Catalog number105325000
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS89-32-7
IUPAC Name5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
Molecular FormulaC10H2O6
InChI KeyANSXAPJVJOKRDJ-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder
Melting point283°C to 287°C
Infrared spectrumConforms
Titration excess NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only